Principaux documents
DWK115300700-B
Wheaton® Microliter Vial
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.11
Matériaux
V-bottom polypropylene bottom
Conditionnement
pkg of 1 packs
Fabricant/nom de marque
WHEATON® 11-5300-700-B
Capacité
900 μL
Application
Chromatography
Caractéristiques et avantages
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.