Principaux documents
DWK07-7000
Wheaton® Microliter μlplates Deep Polypropylene
About This Item
Code UNSPSC :
41106200
Nomenclature NACRES :
NB.11
Matériaux
conical bottom
polypropylene plate
square wells
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
WHEATON® 07-7000
Capacité
2.0 mL
Application
Chromatography
Caractéristiques et avantages
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System uses closures familiar to those who work with standard chromatography vials. Polypropylene ?Lplates have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system permits even GC samples can be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/silicone rubber using a mechanical seal. A variety of insert options let you choose the specifications appropriate for your application. The system combines microplates, glass inserts, Lmats, and caps with chromatography-based septa to offer you secure vessels to maintain even the most volatile samples.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.