Principaux documents
CLS7732
Pyrex® gum stability glass bombs
gum bomb w/ cover, reusable, non-sterile, 1/cs
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass (borosilicate glass code 7740)
cap (Pourout)
Stérilité
non-sterile
Caractéristiques
reusable
Conditionnement
case of 1
pack of 1 ×
Fabricant/nom de marque
Corning 7732
D.E. × H
50 mm × 105 mm
Adéquation
suitable for (the determination of gum stability of gasoline)
Description générale
These PYREX® bombs are for the determination of gum stability of gasoline. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
Informations légales
PYREX is a registered trademark of Corning, Inc.