Key Documents
CLS5000500
Pyrex® wide-mouth Erlenmeyer flask with joint
capacity 500 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5000
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
bottom (baffled)
glass cap
narrow-mouth
Joint
joint ST/NS24/40
Stérilité
non-sterile
Caractéristiques
cap
closure type Standard Taper joint
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 5000-500
Capacité
500 mL
Description générale
  • Pyrex® Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations to show approximate capacity.
  • Large marking spot for labeling.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.