Principaux documents
CLS49602L
Pyrex® round-bottom three neck vertical unequal height flask
capacity 2,000 mL, center joint: ST/NS 45/40
Synonyme(s) :
bottom flask, flask round, flask round bottom
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
three-neck (Vertical)
Joint
center joint: ST/NS45/40
side joint ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper joint
Conditionnement
pack of 1
case of 4
Fabricant/nom de marque
Corning 4960-2L
Capacité
2,000 mL
Description générale
With full length joints.
Informations légales
PYREX is a registered trademark of Corning, Inc.