Principaux documents
CLS4260500
Pyrex® round-bottom boiling flask, short ring neck
capacity 500 mL
Synonyme(s) :
bottom flask, flask round, round flask, Corning® 4260
1 EA
48,10 $
48,10 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
48,10 $
About This Item
Code UNSPSC :
41121804
48,10 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
round bottom flask
short
Taille de l'obturateur
6
Stérilité
non-sterile
Caractéristiques
closure type Ring
Conditionnement
case of 1
Fabricant/nom de marque
Corning 4260-500
Capacité
500 mL
Taille du col
(1)
Description générale
These round bottom boiling flasks have short ring necks and are designed to be used with rubber stoppers. They are precisely tooled for uniform stopper fit. The reinforced rim increases mechanical strength. These flasks are used in applications requiring rigid stoppering and as wash bottles in routine lab washing. Wall thicknesses are carefully controlled to provide the proper balance between thermal expansion and mechanical strength for greater shock resistance. Their excellent chemical durability results in contamination-free results and long life under the toughest lab conditions.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.