Principaux documents
CLS36060350F
Pyrex® Buchner funnel with fritted disc
capacity 350 mL, porosity 4-5.5 μm
Synonyme(s) :
Corning® 36060
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 36060-350F
Capacité
350 mL
Diam. du disque
80 mm
Dimension de pores
4-5.5 μm porosity
Description générale
Especially useful where filter paper would be attacked by filtered material. Disc should not be subjected to pressure differential in excess of 1 atm. Top edge is beaded. Granular frit.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.