Documents
BR90462
BRAND® sampling dipper with spout
capacity 1000 mL, D × H 105 mm × 155 mm
Synonyme(s) :
PTFE, Sampling dippers
About This Item
Code UNSPSC :
41104118
Nomenclature NACRES :
NB.51
Matériaux
PTFE
Fabricant/nom de marque
BRAND 90462
P × H
105 mm × 155 mm
Capacité
1000 mL
Description générale
With reinforced rim and spout. PTFE handle with steel core.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG