Principaux documents
BR90252
BRAND® ETFE beaker with spout, low form
volume 400 mL, with graduation
Synonyme(s) :
Beakers, Plastics
1 EA
103,00 $
103,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
103,00 $
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
103,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
ETFE
Caractéristiques
with graduation
Fabricant/nom de marque
BRAND 90252
Volume
400 mL
Description générale
Made from high-quality ETFE (ethylene-tetrafluoroethylene), this translucent beaker offers excellent chemical resistance, making it ideal for handling various substances in laboratory settings. The low form provides stability and ease of use, allowing for efficient mixing and pouring of liquids. Clearly marked graduations facilitate accurate measurement of liquids, ensuring precision in experiments and formulations. The beaded rim enhances strength and provides a comfortable grip, making handling safer and more efficient.
Autres remarques
Browse all plastic beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG