Principaux documents
BR87604
BRAND® PP beaker with spout, low form
volume 25 mL
Synonyme(s) :
Beakers, Plastics
12 EA
43,60 $
43,60 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
12 EA
43,60 $
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
43,60 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
Description
ISO 7056
Caractéristiques
graduations
beaded rim and spout
Fabricant/nom de marque
BRAND 87604
Volume
25 mL
Description générale
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Autres remarques
Browse all plastic beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG