BR474743
BRAND® microscope slide
ground edges
Synonyme(s) :
Microscope slides, Microscopy
507,00 $
About This Item
507,00 $
Produits recommandés
Matériaux
soda-lime glass
Caractéristiques
ground edges
Fabricant/nom de marque
BRAND 474743
Taille
75 mm × 25 mm × 1 mm
Description générale
- Made from clear soda-lime glass, hydrolytic class 3.
- Ground edges are recommended to reduce the risk of injury.
- Thickness approx.: 1 mm