BR470060
BRAND® cover glass
L × W 24 mm × 24 mm, square
Synonyme(s) :
Cover glasses, Microscopy
231,00 $
About This Item
231,00 $
Produits recommandés
Matériaux
borosilicate glass
square
Fabricant/nom de marque
BRAND 470060
L × l
24 mm × 24 mm
Épaisseur
0.13-0.17 mm , No. 1
Catégories apparentées
Description générale
Caractéristiques et avantages
- BRAND® borosilicate cover glass offers superior clarity and excellent chemical resistance.
- Cover glasses for microscope slides are clean, dust/grease free and distortion-free.
- Full automatic production guarantees "ready-for-use" quality.
- Flatness within ± 3 μm.