Principaux documents
BR4640240
BRAND® Dispensette® S Trace Analysis Analog-adjustable bottle-top dispenser
tantalum spring, volume range 1-10 mL, without recirculation valve
Synonyme(s) :
bottle top dispensers
1 EA
1 970,00 $
1 970,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 970,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
1 970,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
incremental 0.2 mL
without recirculation valve
Fabricant/nom de marque
BRAND 4640240
Gamme de volumes
1-10 mL
Raccords
45 mm
Catégories apparentées
Description générale
Dispensette S Trace Analysis is specialized for dispensing high-purity acids, bases and H2O2.
Also suitable for dispensing of hydrofluoric acid when using platinum-iridium valve spring.
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.