Principaux documents
BR4630361
BRAND® Dispensette® S Organic, Digital bottle-top dispenser
volume range 5-50 mL, with recirculation valve
Synonyme(s) :
bottle top dispensers
1 EA
1 420,00 $
1 420,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 420,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
1 420,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
incremental 0.2 mL
DE-M marking
with recirculation valve
Fabricant/nom de marque
BRAND 4630361
Gamme de volumes
5-50 mL
Raccords
(GL 32-33)
GL 38
S 40
Catégories apparentées
Description générale
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.