Principaux documents
BR4600100
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.1-1 mL, without recirculation valve
Synonyme(s) :
bottle top dispensers
1 EA
789,00 $
789,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
789,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
789,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
incremental 0.02 mL
DE-M marking
without recirculation valve
Fabricant/nom de marque
BRAND 4600100
Gamme de volumes
0.1-1 mL
Raccords
(40 mm)
(GL 24-25)
GL 32
GL 38
S 40
Description générale
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.