Documents
BR43220
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 25 mL, borosilicate glass
Synonyme(s) :
Gay-Lussac
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.71
Matériaux
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
Caractéristiques
with cap
Conditionnement
pack of 2 ea
Fabricant/nom de marque
BRAND 43220
Capacité
25 mL
Description générale
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocoles
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.