Principaux documents
BR28205
BRAND® cotton roving
degreases cotton wool, density - Approx 1.3g per m
Synonyme(s) :
BLAUBRAND®, ETERNA, Graduated Pipettes, Pipettes
1 KG
205,00 $
205,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 KG
205,00 $
About This Item
Code UNSPSC :
42141501
Nomenclature NACRES :
NB.04
205,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
degreases cotton wool
Fabricant/nom de marque
BRAND 28205
Description générale
Cotton plugs may prolong delivery time and may therefore influence the accuracy of measurement. In tests using pipettes that have had cotton plugs inserted by a cotton plugging machine, the results were within the indicated error limits.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG