Principaux documents
BR156650
BRAND® tray stackable
size 840 x 645 x 160 mm
Synonyme(s) :
Photographoc trays, Trays
1 EA
207,00 $
207,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
207,00 $
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
207,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
white
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 156650
Taille
840 x 645 x 160 mm
Catégories apparentées
Description générale
The BRAND® stackable tray is made from durable white polypropylene (PP), designed for versatile use in laboratory and industrial settings. Its ribbed bottom enhances stability, preventing contents from shifting during transport or storage. The reinforced rim adds strength, while the slightly sloping walls facilitate easy access and pouring. This stackable design allows for efficient storage, maximizing space when not in use. Ideal for organizing and handling various materials, this tray combines practicality with durability, making it an essential tool for any workspace.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG