Documents
BR144190
BRAND® atomizer, PE-HD, white
capacity 850 mL
Synonyme(s) :
Atomizer, Bottles, Plastics
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
BRAND 144190
Capacité
850 mL
Description générale
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG