Principaux documents
BAF187601150
Scienceware® POXYGRID® test-tube rack
Holds 60 x 15-16 mm tubes, blue
Synonyme(s) :
tube holder
1 EA
98,80 $
98,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
98,80 $
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
98,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue
epoxy (coated steel)
Caractéristiques
no. of holes 60
Holds 60 x 15-16 mm tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18760-1150
L × l × H
30.5 cm × 21 cm × 6.4 cm
Description générale
Welded steel wire is coated with heat-cured, thermosetting epoxy to protect from rust and corrosion. The coating is unaffected by most organic solvents, weak acids and alkalis, and has excellent abrasion resistance and adhesion to steel substrate.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Informations légales
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP