MilliporeSigma
HomeCell SignalingTissue Dissociation Guide: Collagenase, Dispase, and Liberase Enzyme Types
Tissue Dissociation Guide: Collagenase, Dispase, and Liberase Enzyme Types
General Information about our Tissue Dissociation Enzymes
Collagenases are, enzymes that break down the native collagen that holds animal tissues together and, are made by a variety of microorganisms and by many different animal cells1. The most potent collagenase is the "crude" collagenase secreted by the anaerobic bacteria Clostridium histolyticum. We originally adopted the 1953 fermentation and purification process described by MacLennan, Mandl and Howes2, but eventually improved upon for higher activity products. “Crude” collagenase refers to the fact that the material is a mixture of several different enzymes besides collagenase that act together to break down tissue. It is now known that two forms of the collagenase enzyme are present3,4. With a few exceptions different commercial collagenase are all made from C. histolyticum, or are recombinant versions where Escherichia coli expresses a gene cloned from C. histolyticum.
Description of our Collagenase Enzymes
The different Collagenase products in the tables below were developed by because they each digest some types of tissue (muscle, pancreas, heart, adipose) better than others. In addition to meeting enzyme activity specifications, every lot of our collagenase products must pass digestion tests with various tissues from rats. Products that are also described as “cell culture tested” have undergone additional testing with mammalian cell lines to verify that they are not cytotoxic.
Sterile-filtered (0.2 mm) versions prepared from some of the more popular collagenase products are also listed below.
Our purified collagenase products have only trace amounts of caseinase (proteolytic) or clostripain activities. The purified Type VII Collagenase is also offered in Cell Culture Tested and sterile-filtered versions.
Collagenase Enzyme Products – Crude Collagenase, Chromatographically Purified, Collagenase with Proteolytic Activity Inhibitor, and blends
Crude Collagenase for General Use
ProductCommentsFALGPAaCDUsb
C0130For general use0.25 – 1.0> 125
C1639, Type I-SSterile-filtered version of C01300.25 – 1.0> 125
C9891, Type IASecond version of Type I for general use0.5 – 2.0> 125
C2674, Type IACell culture tested version of C98910.5 – 2.0> 125
C5894, Type IA-SSterile-filtered version of C98910.5 – 2.0> 125
C9722, Type IA-SCell culture tested and sterile-filtered version of C98910.5 – 2.0> 125
a, b FALGPA and Collagenase Digestive Units (CDUs) both given as Units per mg/solid
Crude Collagenase, Use-Tested
ProductCommentsFALGPA aCDUs b
C6885, Type IITested as suitable for digestion and release of fat (adipose) cells with
undamaged cell-surface receptors for insulin5,6
0.5 – 2.0> 125
C1764, Type II-SSterile-filtered version of C68850.5 – 2.0> 125
C5138, Type IVTested as suitable for digestion and release of viable rat liver cells (hepatocytes)70.5 – 2.0> 125
C1889, Type IV-SSterile-filtered version of C51380.5 – 2.0> 125
C2139, Type VIIIPassed both the adipose tissue test (see C6885)5, 6 and
the hepatic tissue test (see C5138)7
0.5 – 2.0> 125
C9263, Type VTested as suitable for digestion and release of islet of
Langerhans (insulin-producing cells) from rat pancreas8,9
1.0 – 3.0> 125
C2014, Type V-SSterile-filtered version of C92631.0 – 3.0> 125
C7657, Type XIHas highest collagenase activity - tested as suitable for
digestion and release of islet of Langerhans (insulin-producing cells) from rat pancreas8,9
2.0 – 5.0> 1,200
C9407, Type XICell culture tested version of C76572.0 – 5.0> 1,200
C4785, Type XI-SSterile-filtered version of C76572.0 – 5.0> 1,200
C9697, Type XI-SCell culture tested and sterile-filtered version of C76572.0 – 5.0> 1,200
a, b FALGPA and Collagenase Digestive Units (CDUs) both given as Units per mg/solid
Chromatographically Purified Collagenase
ProductCommentsFALGPAaCDUsb
C0255, Type IIIPurified collagenase that has trace amounts of protease activity4 – 12≥ 250 
C0773, Type VIIPurified collagenase substantially free of protease and Clostripain4 – 121,000 - 3,000
C2799, Type VIICell culture tested version of C07734 – 121,000 - 3,000
C2399, Type VII-SSterile-filtered version of C07734 – 121,000 - 3,000
C9572, Type VII-SCell culture tested and sterile-filtered version of C07734 – 121,000 - 3,000
a, b FALGPA and Collagenase Digestive Units (CDUs) both given as Units per mg/solid
Crude Collagenase with Proteolytic Activity Inhibitor
ProductCommentsFALGPA aCDUs b
C6079, Type SBlend of Type XI Collagenase with tryptic protease inhibitor.
Tested as suitable for digestion and release of islet of
Langerhans (insulin-producing cells) from rat pancreas8, 9
2 – 5Minimum of 1,000
a, b FALGPA and Collagenase Digestive Units (CDUs) both given as Units per mg/solid
Collagenase Blends™
These products were developed to allow more lot-to-lot reproducibility in collagenase digestions. The proportion of purified collagenase (Blend F) to purified clostridial neutral protease is varied in Blends H and L to provide researchers with a range of digestion options.
ProductCommentsFALGPAaProteasec
C7926, Blend FPredominantly true collagenases. Because FALGPA activity is high compared to
CDU it is assumed that the Type II form of collagenase is mostly present
Min. 2.0< 10
C8051, Blend HSome neutral protease activity included with the collagenase activityMin. 1.0Min. 10
C8176, Blend LMore neutral protease activity is included with true collagenase< 1.0Min. 40
a, c FALGPA and Collagenase Digestive Units (CDUs) both given as Units per mg/solid
Collagenase and Liberase® Enzymes from Roche
Collagenase enzymes from Roche are intuitively designed and trusted to deliver consistent performance and reproducible results for your routine and critical applications. Collagenase is suitable for preparation of cells from many types of tissue, such as hepatocytes, adipocytes, pancreatic islets, epithelial cells, muscle cells, and endothelial cells. However, suitability of each lot of the enzyme for disruption of a tissue should be determined empirically. Additionally, our Liberase® enzyme technology combines highly purified collagenase I and II enzymes with Dispase® or Thermolysin to facilitate the dissociation of a broad range of tissue types.
Liberase Research Grade Application Guide
Tissue TypeTLTMTHDLDH
Liver +  +
Kidney + ++ 
Skin   +++
Heart ++ +
Pancreas (Islets)++  + 
Lymph Node*+++   
Spleen*++    
Lung*++++   
Ovarian Tissue* ++   
Adipose Tissue* ++   
Cornea* ++   
*Data based on publications
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Dispase Enzymes
Along with enzymes available from Roche, we are committed to providing tissue dissociation reagents that are highly purified and dependable for your specific application needs. A gentle enzyme that does not damage cell membranes, dispase is suitable for the separation of diverse tissues and cells that are grown in vitro and to prevent cell clumping for suspension cultures.
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Collagenase Assays
The Type I and Type II forms of the purified collagenase enzymes differ in their specificities and relative activities on native collagen and synthetic substrates. These two collagenases can be mostly distinguished by their preference for one of the two different substrates used in our assays. The Collagenase Digestive Unit (CDU) assay10,11 measures predominantly the Collagenase I activity, which cleaves two of the three helical chains in the long, undenatured collagen protein. Collagenase II activity is measured by this enzyme’s ability to cut a short synthetic peptide, N-[3-(2-Furyl)acryloyl)]-Leu-Gly-Pro-Ala (FALGPA, see Product No. F5135), in a second collagenase digestive assay12,13. Purified preparations of either Collagenase I or II have been shown to be less effective at digesting various types of collagen or mammalian tissue when compared to a mixture of both forms of this enzyme. A purified collagenase containing only the Collagenase I and II forms of this enzyme is less effective at digesting tissue than the whole crude collagenase or combinations of the purified collagenase and various proteases. Obviously, the combination of true collagenase and the different native proteases, Clostripain and aminopeptidases that have evolved in nature assist each other in digesting the collagen in different animal tissues. For tissue digestions the crude collagenase products have always been the most effective. Some researchers have tried mixtures of chromatographically purified collagenase with a protease such as trypsin or subtilisin to digest tissue.
In addition to the CDU and FALGPA assays for Collagenase activities, we test each product lot for Caseinase14,15, Clostripain and Tryptic activities to look at the proteolytic enzymatic activities in the collagenase products. The Caseinase assay is the most important of the three for measuring the proteolytic activity that assists the digestion of animal tissue. Because the Clostripain present in crude collagenase must be reduced (e.g. by treatment with Dithiothreitol) in order to be active this enzyme probably contributes little to the tissue dissociation process in the laboratory. It is monitored because some researchers have reported that Clostripain may be damaging or toxic.
Many collagenase products that meet enzymatic specifications are also use-tested with various tissues obtained from rats. Type II (C6885C1764) and Type VIII (C2139) collagenase lots are tested for the ability to release adipose (fat) cells from rat epididymal fat pads5. Fat cells are then screened for metabolic activity by measuring glucose oxidation rates with and without insulin addition. Type IV (C5138, C1889) and Type VIII (C2139) lots have been tested for the ability to release viable cells from rat liver7. Type V (C9263, C2014), Type XI (C7657, C4785, C9407, and C9697) and Type S (C6079) Collagenase lots must release intact islets of Langerhans from rat pancreas to pass their product test8.
Collagenase Tissue Digestion/Dissocation Troubleshooting and References
Based on our own R&D and from discussions with customers it is clear that the way a particular tissue is dissected and prepared has a significant effect on the speed and efficiency of any tissue digestion-dissociation with collagenase. Differences in the ages of the tissue donors can also be a major source of variation over time. Make sure that calcium ions are present in the digestion buffers at 5 mM. Chelating agents EGTA and EDTA can severely inhibit Collagenase activity by removing Calcium ions required for enzyme stability and activity. β-mercaptoethanol16, cysteine16 and 8-hydroxyquinoline-5-sulfonate16 ;are other inhibiting substances. A new lot of Collagenase with higher specific activity could cause excessive cell death at an established concentration. In that case use less collagenase and/or add BSA or serum (up to 0.5% and 5-10% respectively) to stabilize the cells to further digestion.
ProblemCauseSolutionReference
Digestion is poorInactive Enzymes
Inadequate Ca++
Insufficient Enzymes
Store Collagenase cold and dry.
Store collagenase solution in frozen aliquots.
Include 5 mM Ca++ in collagenase solution.
Use more collagenase. Try our Blends with more protease activity.
6
7
Released Cells are TrappedDNA released from broken cells
Inadequate Non-protein gums
Reduce agitation
Add DNAse**
Flush tissue well before digesting it***
Use no Mg++ in digestion solution
Add hyaluronidase with enzymes**
7
9
7
7
18
Cells Are KilledExcess protease
pH changes
Too little oxygen
Reduce exposure to proteases*
Add albumin or heated serum
Use buffers (e.g. HEPES) instead of HCO-3.
Check and readjust pH often
Aerate digestion solution (air or O2)
Digest faster by using more enzymes
7
19
Cell Yield is LowEnzyme balance
Adhesion factors
Use more Collagenase*
Include elastase with Collagenase**
Perfuse tissue first to remove Ca++***
20
7
Many Cells are DamagedExcessive Protease
Physical damage
Use less protease*
Add albumin or heated serum
Handle tissue and cells very gently
21
7
New Lot Doesn’t workLot variationUse fractionated collagenase “blends” (Product No. C7926C8051 or C8176)* 
* The separately prepared Collagenase and protease enzymes in the “Blend” products (No. C7926C8051 or C8176) give reproducible control of how much of each is used.
** DNAse will be inactivated by the shear of excessive stirring, and added enzymes may be digested by the neutral protease present in the Collagenase.
*** Use EGTA (or EDTA) to remove Ca++ and flush away microorganisms, then wash tissue with buffer to remove the chelating agent. Do not add EGTA or EDTA to the enzyme solutions!
References
1.
Harper E. 1980. Collagenases. Annu. Rev. Biochem.. 49(1):1063-1078. https://doi.org/10.1146/annurev.bi.49.070180.005215
2.
MacLennan JD, Mandl I, Howes EL. 1953. BACTERIAL DIGESTION OF COLLAGEN 1. J. Clin. Invest.. 32(12):1317-1322. https://doi.org/10.1172/jci102860
3.
Bond MD, Van Wart HE. 1984. Characterization of the individual collagenases from Clostridium histolyticum. Biochemistry. 23(13):3085-3091. https://doi.org/10.1021/bi00308a036
4.
Matsushita O, Jung C, Katayama S, Minami J, Takahashi Y, Okabe A. 1999. Gene Duplication and Multiplicity of Collagenases in Clostridium histolyticum. J. Bacteriol.. 181(3):923-933. https://doi.org/10.1128/jb.181.3.923-933.1999
5.
Rodbell M. 1964. Metabolism of Isolated Fat Cells. Journal of Biological Chemistry. 239(2):375-380. https://doi.org/10.1016/s0021-9258(18)51687-2
6.
Fain JN. 1975. [53] Isolation of free brown and white fat cells.555-561. https://doi.org/10.1016/0076-6879(75)35184-7
7.
Seglen PO. 1976. Chapter 4 Preparation of Isolated Rat Liver Cells.29-83. https://doi.org/10.1016/s0091-679x(08)61797-5
8.
Lacy PE, Kostianovsky M. 1967. Method for the Isolation of Intact Islets of Langerhans from the Rat Pancreas. Diabetes. 16(1):35-39. https://doi.org/10.2337/diab.16.1.35
9.
Buitrago A, Gylfe E, Henriksson C, Pertoft H. 1977. Rapid isolation of pancreatic islets from collagenase digested pancreas by sedimentation through percoll? at unit gravity. Biochemical and Biophysical Research Communications. 79(3):823-828. https://doi.org/10.1016/0006-291x(77)91185-8
10.
Moore S, Stein WH. 1948. PHOTOMETRIC NINHYDRIN METHOD FOR USE IN THE CHROMATOGRAPHY OF AMINO ACIDS. Journal of Biological Chemistry. 176(1):367-388. https://doi.org/10.1016/s0021-9258(18)51034-6
11.
“Enzymatic Assay of Collagenase. Collagen Digestion Assay”, our quality control test procedure..
12.
Van Wart HE, Steinbrink D. 1981. A continuous spectrophotometric assay for Clostridium histolyticum collagenase. Analytical Biochemistry. 113(2):356-365. https://doi.org/10.1016/0003-2697(81)90089-0
13.
“Enzymatic Assay of Collagenase Using FALGPA as the Substrate”, our quality control test procedure..
14.
Anson M, Gen. Physiol. J. The Estimitation of Pepsin, Trypsin, Papain and Cathepsin with Hemoglobin. 22, 79 (1938).
15.
“Enzymatic Assay of Caseinase (Collagenase Products)”, our quality control test procedure..
16.
Seifter S, Gallop PM, Klein L, Meilman E. 1959. Studies on Collagen. Journal of Biological Chemistry. 234(2):285-293. https://doi.org/10.1016/s0021-9258(18)70290-1
17.
Berry MN, Friend DS. 1969. HIGH-YIELD PREPARATION OF ISOLATED RAT LIVER PARENCHYMAL CELLS. 43(3):506-520. https://doi.org/10.1083/jcb.43.3.506
18.
Bellemann P, Gebhardt R, Mecke D. 1977. An improved method for the isolation of hepatocytes from liver slices. Analytical Biochemistry. 81(2):408-415. https://doi.org/10.1016/0003-2697(77)90711-4
19.
Ives HE, Schultz GS, Galardy RE, Jamieson JD. 1978. Preparation of functional smooth muscle cells from the rabbit aorta.. 148(5):1400-1413. https://doi.org/10.1084/jem.148.5.1400
20.
Fain JN, Loken SC. 1969. Response of Trypsin-treated Brown and White Fat Cells to Hormones. Journal of Biological Chemistry. 244(13):3500-3506. https://doi.org/10.1016/s0021-9258(18)83400-7
21.
Berry, M, Edwards, Aa, Barritt, G. 1991. Isolated Hepatocytes; Preparation, Properties and Applications. Elsevier. .