MilliporeSigma
HPLC Analysis of Conjugated Estrogens on Discovery® HS C18
HPLC Analysis of Conjugated Estrogens on Discovery® HS C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
β-Estradiol 3-(β-D-glucuronide) sodium salt
β-Estradiol 3-(β-D-glucuronide) 17-sulfate dipotassium salt
β-Estradiol 3-sulfate sodium salt
≥93%
Estrone 3-(β-D-glucuronide) sodium salt
Estriol
meets USP testing specifications
β-Estradiol 3,17-disulfate dipotassium salt
≥95%
α-Estradiol
powder, ≥98% (TLC)
β-Estradiol 17-(β-D-glucuronide) sodium salt
Estriol 3-sulfate sodium salt
≥98% (TLC)
Estrone
meets USP testing specifications
Estrone 3-sulfate sodium salt
contains ~35% Tris as stabilizer
17β-Estradiol solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Estriol solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Discovery HS C18, 15 cm × 4.6 mm I.D., 5 μm particles (568522-U)
mobile phase
[A] 10 mM potassium phosphate (pH 7.01 unadjusted); [B] acetonitrile; [C] methanol
gradient
80% A, 15% B, 5% C, held for 5 min; to 40% A, 45% B, 15% C in 20 min; held for 5 min
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
10 μL
sample
10 μg/mL each in 60/40, water:acetonitrile
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany