Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E1274

Sigma-Aldrich

Estrone

meets USP testing specifications

Synonym(s):

1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
1915077
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
meets USP testing specifications

mp

258-260 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annamaria Lilienkampf et al.
Journal of medicinal chemistry, 52(21), 6660-6671 (2009-10-15)
Many breast tumors are hormone-dependent, and estrogens, especially estradiol (E2), have a pivotal role in their growth and development. 17beta-Hydroxysteroid dehydrogenase type 1 (17beta-HSD1) is a key enzyme in the biosynthesis of female sex steroids, catalyzing the NADPH-dependent reduction of
Emma M Ferguson et al.
Water research, 47(4), 1604-1615 (2013-01-12)
Estuaries are often the final repositories for aquatic pollutants but how estuarine hydrology influences the availability of marine- and freshwater-derived pollutants is not well understood, particularly for micro-pollutants such as endocrine disrupting chemicals. To address this knowledge gap, this study
Juilee Rege et al.
The Journal of clinical endocrinology and metabolism, 98(3), 1182-1188 (2013-02-07)
A broad analysis of adrenal gland-derived 19-carbon (C19) steroids has not been reported. This is the first study that uses liquid chromatography-tandem mass spectrometry to quantify 9 C19 steroids (androgens and their precursors), estrone, and estradiol in the adrenal vein
Yannick Laplante et al.
Bioorganic & medicinal chemistry, 16(4), 1849-1860 (2007-11-24)
Estrogens play an important role in the development of breast cancer. Inhibiting 17beta-hydroxysteroid dehydrogenase type 1 (17beta-HSD1)--the enzyme responsible for the last step in the biosynthesis of the most potent estrogen, estradiol (E2)--would thus allow hindering the growth of estrogen-sensitive
Shanshan Zhao et al.
Breast cancer research : BCR, 16(2), R30-R30 (2014-03-29)
Paradoxically, a breast cancer risk reduction with conjugated equine estrogens (CEE) and a risk elevation with CEE plus medroxyprogesterone acetate (CEE + MPA) were observed in the Women's Health Initiative (WHI) randomized controlled trials. The effects of hormone therapy on serum sex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service