MilliporeSigma
  • Production of monoclonal antibodies and development of a quantitative immuno-polymerase chain reaction assay to detect and quantify recombinant Glutathione S-transferase.
Production of monoclonal antibodies and development of a quantitative immuno-polymerase chain reaction assay to detect and quantify recombinant Glutathione S-transferase.
Protein expression and purification (2017-05-02)
J E Abud, E H Luque, J G Ramos, H A Rodriguez
ABSTRACT
GST-tagged proteins are important tools for the production of recombinant proteins. Removal of GST tag from its fusion protein, frequently by harsh chemical treatments or proteolytic methods, is often required. Thus, the monitoring of the proteins in tag-free form requires a significant effort to determine the remnants of GST during purification process. In the present study, we developed both a conventional enzyme-linked immunosorbent assay (ELISA) and an immuno-polymerase chain reaction (IPCR) assay, both specific for detection of recombinant GST (rGST). rGST was expressed in Escherichia coli JM109, using a pGEX4T-3 vector, and several anti-rGST monoclonal antibodies were generated using hybridoma technology. Two of these were rationally selected as capture and detection antibodies, allowing the development of a sandwich ELISA with a limit of detection (LOD) of 0.01 μg/ml. To develop the rGST-IPCR assay, we selected "Universal-IPCR" format, comprising the biotin-avidin binding as the coupling system. In addition, the rGST-IPCR was developed in standard PCR tubes, and the surface adsorption of antibodies on PCR tubes, the optimal neutravidin concentrations, the generation of a reporter DNA and the concentration effect were studied and determined. Under optimized assay conditions, the rGST-IPCR assay provided a 100-fold increase in the LOD as well as an expanded working range, in comparison with rGST-ELISA. The proposed method exhibited great potentiality for application in several fields in which measurement of very low levels of GST is necessary, and might provide a model for other IPCR assays.
Sigma-Aldrich
Human KIT / Mast/Stem Cell Growth Factor Receptor Kit ELISA Kit