Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0731

Sigma-Aldrich

Human KIT / Mast/Stem Cell Growth Factor Receptor Kit ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KIT(3815)

General description

The antibody pair provided in this kit recognizes human Mast/Stem Cell Growth Factor Receptor Kit.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atta Muhammad Memon et al.
Viral immunology, 30(4), 264-270 (2017-04-18)
Porcine rotavirus-A (PoRVA) is one of the common causes of mild to severe dehydrating diarrhea, leading to losses in weaning and postweaning piglets. A rapid, highly specific, and sensitive antigen-capture enzyme-linked immunosorbent assay (AC-ELISA) was developed for detection of PoRVA
Bingyang Lyu et al.
International journal of environmental research and public health, 16(24) (2019-12-11)
Forest therapy is a fast-growing treatment approach, as it has the potential to alleviate stressful life events and to improve psychological well-being and physical health. Bamboo forests are widespread in southwestern China. Nevertheless, a knowledge gap on the specific health
Małgorzata Pomorska-Mól et al.
BMC veterinary research, 13(1), 376-376 (2017-12-06)
Respiratory co-infections are important factor affecting the profitability of pigs production. Swine influenza virus (SIV) may predispose to secondary infection. Haemophilus parasuis (Hps) can be a primary pathogen or be associated with other pathogens such as SIV. To date, little
Michael Kruse et al.
Molecular nutrition & food research, 64(21), e2000419-e2000419 (2020-09-14)
Effective treatment for obesity associated non-alcoholic fatty liver disease (NAFLD) is limited. Dietary supplementation of n-3 polyunsaturated fatty acids, specifically alpha linolenic acid (ALA), can resolve intrahepatic lipid content (IHL). This study investigates the effect of daily supplementation of either
Małgorzata Pomorska-Mól et al.
Journal of veterinary internal medicine, 34(5), 1903-1913 (2020-07-04)
Simultaneous viral infections exhibit the phenomenon of viral interference, but understanding of the effect of one virus on another is limited. Evaluate and compare clinical characteristics, immune and acute phase response, viral shedding and viral load in pigs singly and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service