MilliporeSigma
  • Regulation of subcellular distribution and oncogenic potential of nucleophosmin by plakoglobin.
Regulation of subcellular distribution and oncogenic potential of nucleophosmin by plakoglobin.
Oncogenesis (2012-01-01)
L Lam, Z Aktary, M Bishay, C Werkman, C-Y Kuo, M Heacock, N Srivastava, J R Mackey, M Pasdar
ABSTRACT
Nucleophosmin (NPM) is a nucleolar phosphoprotein that is involved in many cellular processes and has both oncogenic and growth suppressing activities. NPM is localized primarily in nucleoli but shuttles between the nucleus and the cytoplasm, and sustained cytoplasmic distribution contributes to its tumor promoting activities. Plakoglobin (PG, γ-catenin) is a homolog of β-catenin with dual adhesive and signaling functions. These proteins interact with cadherins and mediate adhesion, while their signaling activities are regulated by association with various intracellular partners. Despite these similarities, β-catenin has a well-defined oncogenic activity, whereas PG acts as a tumor/metastasis suppressor through unknown mechanisms. Comparison of the proteomic profiles of carcinoma cell lines with low- or no PG expression with their PG-expressing transfectants has identified NPM as being upregulated upon PG expression. Here, we examined NPM subcellular distribution and in vitro tumorigenesis/metastasis in the highly invasive and very low PG expressing MDA-MB-231 (MDA-231) breast cancer cells and their transfectants expressing increased PG (MDA-231-PG) or NPM shRNA (MDA-231-NPM-KD) or both (MDA-231-NPM-KD+PG). Increased PG expression increased the levels of nucleolar NPM and coimmunoprecipitation studies showed that NPM interacts with PG. PG expression or NPM knockdown decreased the growth rate of MDA-231 cells substantially and this reduction was decreased further in MDA-231-NPM-KD+PG cells. In in vitro tumorigenesis/metastasis assays, MDA-231-PG cells showed substantially lower and MDA-231-NPM-KD cells substantially higher invasiveness relative to the MDA-231 parental cells, and the co-expression of PG and NPM shRNA led to even further reduction of the invasiveness of MDA-231-PG cells. Furthermore, examination of the levels and localization of PG and NPM in primary biopsies of metastatic infiltrating ductal carcinomas revealed coordinated expression of PG and NPM. Together, the data suggest that PG may regulate NPM subcellular distribution, which may potentially change the function of the NPM protein from oncogenic to tumor suppression.
Sigma-Aldrich
MISSION® esiRNA, targeting human NPM1