MilliporeSigma
  • Hyaluronan up-regulation is linked to renal dysfunction and hearing loss induced by silver nanoparticles.
Hyaluronan up-regulation is linked to renal dysfunction and hearing loss induced by silver nanoparticles.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery (2014-08-02)
Hao Feng, Ilmari Pyykkö, Jing Zou
ABSTRACT
Increased application of silver nanoparticles (AgNPs) has raised concerns on their potential adverse effects on human health. However, the precise toxicological mechanisms are not known in detail. The current study hypothesized that AgNPs induced glycosaminoglycan accumulation in the basement membrane that associated with the up-regulation of its component hyaluronic acid, known as a hydrophilic molecule of binding and retaining water, and caused toxicities in the kidney and cochlea. Rats administered AgNPs through either intravenous or intratympanic injection were observed at different time points after exposure. The concentrations of creatinine and urea in the serum were elevated remarkably, and proteins leaked into the urine were increased. A significant hearing loss over a broad range of frequencies was indicated. AgNP exposure induced glycosaminoglycan accumulation and hyaluronic acid up-regulation in the basement membrane. Abundant apoptotic cell death was demonstrated in the AgNP-exposed organs. Our results suggested that glycosaminoglycan accumulation associated with the up-regulation of hyaluronic acid was involved in the toxicities of kidney and cochlea caused by AgNPs.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)