MilliporeSigma
  • Prevalence and Characterization of High Histamine-Producing Bacteria in Gulf of Mexico Fish Species.
Prevalence and Characterization of High Histamine-Producing Bacteria in Gulf of Mexico Fish Species.
Journal of food protection (2015-07-22)
Kristin Bjornsdottir-Butler, John C Bowers, Ronald A Benner
ABSTRACT
Recent developments in detection and enumeration of histamine-producing bacteria (HPB) have created powerful molecular-based tools to better understand the presence of spoilage bacteria and conditions, resulting in increased risk of scombrotoxin fish poisoning. We examined 235 scombrotoxin-forming fish from the Gulf of Mexico for the presence of high HPB. Photobacterium damselae subsp. damselae was the most prevalent HPB (49%), followed by Morganella morganii (14%), Enterobacter aerogenes (4%), and Raoultella planticola (3%). The growth characteristics and histamine production capabilities of the two most prevalent HPB were further examined. M. morganii and P. damselae had optimum growth at 35°C and 30 to 35°C and 0 to 2% and 1 to 3% NaCl, respectively. P. damselae produced significantly (P < 0.001) higher histamine than M. morganii in inoculated mahimahi and Spanish mackerel incubated at 30°C for 24 h, but histamine production was not significantly different between the two HPB in inoculated tuna, possibly due to differences in muscle composition and salt content. Results in this study showed that P. damselae was the most prevalent high HPB in Gulf of Mexico fish. In addition, previously reported results using the traditional Niven's method may underreport the prevalence of P. damselae. Molecular-based methods should be used in addition to culture-based methods to enhance detection and enumeration of HPB.
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O