MilliporeSigma
  • Development of tyrosinase biosensor based on quantum dots/chitosan nanocomposite for detection of phenolic compounds.
Development of tyrosinase biosensor based on quantum dots/chitosan nanocomposite for detection of phenolic compounds.
Analytical biochemistry (2015-07-15)
En Han, Yi Yang, Zheng He, Jianrong Cai, Xinai Zhang, Xiaoya Dong
ABSTRACT
A sensitive and simple amperometric biosensor for phenols was developed based on the immobilization of tyrosinase into CdS quantum dots/chitosan nanocomposite matrix. The nanocomposite film with porous nanostructure, excellent hydrophilicity and biocompatibility resulted in high enzyme loading, and the tyrosinase (Tyr) immobilized in this novel matrix retained its activity to a large extent. The CdS quantum dots/chitosan nanocomposite film was characterized by scanning electron microscopy and electrochemical impedance spectroscopy, and the parameters of the various experimental variables for the biosensor were optimized. Under the optimal conditions, the designed biosensor displayed a wide linear response to catechol over a concentration range of 1.0×10(-9) to 2.0×10(-5)M with a high sensitivity of 561±9.7mAM(-1) and a low detection limit down to 0.3 nM at a signal-to-noise ratio of 3. The CdS quantum dots/chitosan nanocomposites could provide a novel matrix for enzyme immobilization to promote the development of biosensing and biocatalysis.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
2-Mercaptopropionic acid, ≥95%, FG
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
m-Cresol, ≥98%, FG
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%