MilliporeSigma
  • A Systematic Study on Manufacturing of Prilled Microgels into Lipids for Oral Protein Delivery.
A Systematic Study on Manufacturing of Prilled Microgels into Lipids for Oral Protein Delivery.
Journal of pharmaceutical sciences (2015-06-26)
Jan Kendall De Kruif, Felipe Varum, Roberto Bravo, Martin Kuentz
ABSTRACT
The development of novel systems with oral protein delivery as ultimate goal represents an important field of pharmaceutics. Prilling of protein-loaded polymeric solutions into lipid-based hardening baths could provide here an attractive formulating technology. As the obtained microgel dispersion can be directly capsule-filled, no drying step is required and thermal drug degradation is avoided. This study aims to find excipient combinations for the novel prilling process and investigate systematically diverse material and process factors. Bovine serum albumin and mono-N-carboxymethyl chitosan were selected as model protein and prilling polymer, respectively. The prilling suitability of 880 formulations was screened with 60 ternary phase diagrams comprising two co-solvents, 10 different glycerides, and three so-called complementary excipients. Preliminary capsule compatibility was tested for one month on 245 formulations in hard and soft capsules with different shell materials. Ternary phase diagrams' center points were used to evaluate morphology, encapsulation efficiency, and protein stability of the prilled microgels. As result, several formulations proved suitable for prilling and compatible for capsule filling. Statistical analysis using partial least square regression revealed significant factors regarding different quality attributes of microgel dispersions. Therefore, an improved understanding was obtained for this promising drug delivery approach.
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, purum p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis