MilliporeSigma
  • Infrared Spectrum and UV-Induced Photochemistry of Matrix-Isolated 5-Hydroxyquinoline.
Infrared Spectrum and UV-Induced Photochemistry of Matrix-Isolated 5-Hydroxyquinoline.
The journal of physical chemistry. A (2015-05-31)
Nihal Kuş, Seda Sagdinc, Rui Fausto
ABSTRACT
The structure, infrared spectrum, and photochemistry of 5-hydroxyquinoline (5HQ) were studied by matrix isolation infrared spectroscopy, complemented by theoretical calculations performed at the DFT(B3LYP)/6-311++G(d,p) level of approximation. According to the calculations, the trans conformer of 5HQ (with the OH group pointing to the opposite direction of the pyridine ring of the molecule) is more stable than the cis form (by ∼8.8 kJ mol(-1)). The main factors determining the relative stability of the two conformers were rationalized through natural bond orbital (NBO) and charge density analyses. The compound was trapped in solid nitrogen at 10 K, and its infrared spectra registered and interpreted, showing the sole presence in the matrix of the more stable trans conformer. Broadband in situ UV irradiations (λ ≥ 288 nm and λ ≥ 235 nm) allowed for the observation of different chemical transformations, which started by excitation to the S1 state of 5HQ, followed by homolytic cleavage of the O-H bond, and subsequent reattachment of the H atom to the 5HQ radical to form quinolin-5(6H)-one and quinolin-5(8H)-one. The first of these two quinolinones was found to convert to open-ring isomeric ketenes, especially when irradiation was performed at higher energy, whereas the second is rather stable under the used experimental conditions. As a whole, the observed photochemistry of matrix-isolated 5HQ closely matches those previously reported for phenol and thiophenol. A detailed mechanistic interpretation for the observed photochemical processes is here proposed, which received support from time-dependent DFT calculations.
Sigma-Aldrich
Thiophenol, ≥99%
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, natural, 97%, FG