MilliporeSigma
  • Autocrine positive regulatory feedback of glucocorticoid secretion: glucocorticoid receptor directly impacts H295R human adrenocortical cell function.
Autocrine positive regulatory feedback of glucocorticoid secretion: glucocorticoid receptor directly impacts H295R human adrenocortical cell function.
Molecular and cellular endocrinology (2014-07-25)
Laetitia Asser, Ségolène Hescot, Say Viengchareun, Brigitte Delemer, Séverine Trabado, Marc Lombès
ABSTRACT
Glucocorticoid receptor (GR), a ubiquitous transcriptional factor, regulates target gene expression upon activation by glucocorticoids, notably cortisol, a corticosteroid hormone synthesized in the adrenal cortex. We thus hypothesized that both GR and cortisol might be involved in the regulation of adrenal physiology and steroidogenesis in an autocrine manner. In a cortisol-secreting human adrenocortical cell line (H295R), the GR-dependent signaling pathway was pharmacologically modulated either by dexamethasone (DEX), a GR agonist or by RU486, a GR antagonist, or was knocked-down by small interfering RNA strategy (SiRNA). We showed that GR activation, elicited by 48 h exposure to DEX, exerts a global positive regulatory effect on adrenal steroidogenesis as revealed by a 1.5- to 2-fold increase in cortisol, 11-deoxycortisol and 17-hydroxyprogesterone secretion associated with a significant enhanced expression of steroidogenesis factors such as StAR, CYP11A1, CYP21A2 and CYP11B1. In sharp contrast, RU486 treatment exerted opposite effects by decreasing both steroid production and expression of these steroidogenic factors. Likewise, GR repression by SiRNA also significantly reduced StAR, CYP11A1, and CYP11B1 mRNA levels. Interestingly, RU486 resulted in a significant CYP21A2 enzymatic blockade as demonstrated by a massive increase in 17-hydroxyprogesterone concentrations in RU486-treated H295R cell supernatants, while cortisol and 11-deoxycortisol secretions were reduced by more than 60%. Consistently, we also demonstrated that metabolic conversion of 17-hydroxyprogesterone into 11-deoxycortisol onto H295R cells was drastically blunted in the presence of RU 486. Finally, steady state levels of MC2R transcripts encoding for the ACTH receptor were significantly induced by DEX, unlikely through a direct GR-mediated transcriptional activation as opposed to CYP11A1 and FKBP5 target genes. These results could account for a higher glucocorticoid-elicited ACTH sensitivity of adrenocortical cells. Our study identifies a positive ultra-short regulatory loop exerted by GR on steroidogenesis in H295R cells, thus supporting a complex intra-adrenal GR-mediated feedback, likely relevant for human adrenocortical pathologies.
Product Description
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology