MilliporeSigma
Stem cell programs are retained in human leukemic lymphoblasts.
Oncogene (2014-06-10)
D Fan, X Zhou, Z Li, Z-Q Li, C Duan, T Liu, F Zhang, Y Huang, Y Zhang, F Gao, Y Guo, R Gupta, G Chen, T Enver, J Tang, D Hong
ABSTRACT
Leukemic lymphoblasts within different immunophenotypic populations possess stem cell properties. However, whether or not the self-renewal program is retained from stem cells or conferred on progenitors by leukemogenic molecules remains unknown. We have addressed the issue in the context of TEL-AML1-associated acute lymphoblastic leukemia (ALL) by profiling a refined program edited from genes essential for self-renewal of hematopoietic stem cells and B-cell development. Bioinformatic analysis shows that ALL populations are loosely clustered and close to the normal population that contains stem and primitive progenitor cells. This finding indicates that immunophenotypes do not reflect maturation stages in ALL and that the self-renewal program may be retained from stem cells. Results of assessing 'first hit' function of TEL-AML1 in different populations of normal cells demonstrate the molecular model. Therefore, the current study shows a leukemogenic scenario of human ALL in which programs of stem cells are sustained in distinct fractions by leukemogenic mutations.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium hydride, in paraffin
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder