MilliporeSigma
  • Dexamethasone treatment alters function of adipocytes from a mesenchymal stromal cell line.
Dexamethasone treatment alters function of adipocytes from a mesenchymal stromal cell line.
Biochemical and biophysical research communications (2014-08-01)
James C Johnston, Azeb Haile, Dongqing Wang, Gabriele Ronnett, Lynne C Jones
ABSTRACT
Osteoporosis and osteonecrosis are associated with corticosteroid treatment, but the pathophysiologies are unclear. We hypothesized that mature adipocytes present within the bone marrow compartment play a key role in the development of both diseases. Adipocytes have recognized regulatory effects on bone viability and healing by releasing signaling molecules called adipokines. Our purpose was to evaluate whether dexamethasone alters adipokine expression in differentiated bone-marrow-derived adipocytes. Adipocytes differentiated from mouse D1 mesenchymal stromal cells were treated with dexamethasone (10(-5), 10(-6), 10(-7), or 10(-8)M) or with diluent alone (controls) for up to 6days. Using real-time polymerase chain reaction and enzyme-linked immunosorbent assay analyses, six key adipokines and the transcription factor HIF-1α were evaluated. Dexamethasone treatment increased PAI-1 protein expression with increased mRNA expression at 4days, while decreasing HIF-1α mRNA expression and protein concentrations. VEGF A mRNA expression was increased at 4days for most dexamethasone concentrations, with minimal changes in protein levels. Dexamethasone increased adiponectin mRNA expression and protein levels at 4 and 6days and decreased leptin, interleukin-6, and tumor necrosis factor α mRNA expression at all time periods. Dexamethasone treatment of bone-marrow-derived adipocytes resulted in detectable changes in mRNA expression and protein levels of adipokines and HIF-1α. The detected adipokine alterations could be important early events in the pathogenesis of steroid-induced osteonecrosis and osteoporosis.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Supelco
Dexamethasone, VETRANAL®, analytical standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source