MilliporeSigma
  • Overexpression of cyclin dependent kinase inhibitor P27/Kip1 increases oligodendrocyte differentiation from induced pluripotent stem cells.
Overexpression of cyclin dependent kinase inhibitor P27/Kip1 increases oligodendrocyte differentiation from induced pluripotent stem cells.
In vitro cellular & developmental biology. Animal (2014-04-26)
Shinpei Tamaki, Yasuhito Tokumoto
ABSTRACT
Cell transplantation therapy with oligodendrocyte precursor cells (OPCs) is a promising and effective treatment for diseases involving demyelination in the central nervous system (CNS). In previous studies, we succeeded in producing O4(+) oligodendrocytes (OLs) from mouse- and human-induced pluripotent stem cells (iPSCs) in vitro; however, the efficiency of differentiation into OLs was lower for iPSCs than that for embryonic stem cells (ESCs). To clarify the cause of this difference, we compared the expression of proteins that contribute to OL differentiation in mouse iPSC-derived cells and in mouse ESC-derived cells. The results showed that the expression levels of cyclin dependent kinase inhibitor P27/Kip1, mitogen-activated protein kinase (MAPK) JNK3, and transcription factor Mash1 were lower in iPSC-derived cells. In contrast, the expression levels of MAPK P38α, P38γ, and thyroid hormone receptor β1 were higher in iPSC-derived cells. We attempted to compensate for the expression changes in P27/Kip1 protein and Mash1 protein in iPSC-derived cells through retrovirus vector-mediated gene expression. Although the overexpression of Mash1 had no effect, the overexpression of P27/Kip1 increased the differentiation efficiency of iPSC-derived cells into O4(+) OLs.
Sigma-Aldrich
Leukemia Inhibitory Factor human, animal component free, recombinant, expressed in rice, 500 μg/mL
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Leukemia Inhibitory Factor human, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution (pH 7.4), suitable for cell culture
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
LIF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
USP
Liothyronine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)