MilliporeSigma
  • Application of the equivalency factor concept to the phototoxicity and -genotoxicity of furocoumarin mixtures.
Application of the equivalency factor concept to the phototoxicity and -genotoxicity of furocoumarin mixtures.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-01)
Nicole Raquet, Dieter Schrenk
ABSTRACT
Furocoumarins (FCs) are natural constituents widely occurring in plants used as food or in phytomedicines, cosmetics, etc. Some FCs exert dermal photo-toxicity and -genotoxicity when combined with UVA irradiation. For a few congeners, skin tumor formation has been described in humans and laboratory animals. Since almost no information is available on the photo-toxic properties of several congeners, we analyzed the photo-cytotoxic, photo-mutagenic, and photo-clastogenic properties in V79 cells for thirteen naturally occurring FCs, and for the coumarin limettin. Furthermore, nine FC mixtures including one mixture based on the FC pattern of an Angelica archangelica extract were tested in the same assays. We found that the concept of relative potency factors for photo-cytotoxic, -mutagenic, and -clastogenicpotencies of FCs, setting the value for 5-methoxypsoralen at 1.00, was applicable to all congeners tested. The concept was used successfully to describe the photo-toxic properties of binary mixtures of 5- and 8-methoxypsoralen. Furthermore, the photo-genotoxic (photo-mutagenic and -clastogenic) properties of complex FC mixtures comprising up to nine different congeners could be predicted. These data suggest that FCs can differ widely in their photo-toxic and photo-genotoxic properties but show relatively strict additivity with respect to their on target-effects when occurring as complex mixtures.
Sigma-Aldrich
5-Methoxypsoralen, 99%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Psoralen, ≥99% (HPLC)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Angelicin
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Supelco
8-Methoxypsoralen, analytical standard
Supelco
Xanthotoxin, analytical standard
Supelco
Bergapten, analytical standard