MilliporeSigma
Effects of vipoxin and its components on HepG2 cells.
Toxicon : official journal of the International Society on Toxinology (2014-12-24)
Jordan Doumanov, Kirilka Mladenova, Tanya Topouzova-Hristova, Stoyanka Stoitsova, Svetla Petrova
ABSTRACT
Snake venom Phospholipases A2 (svPLA2) are among the main toxic venom components with a great impact on different tissues and organs based on their catalytic specificity and a variety of pharmacological effects, whose mechanism is still under debate. The main toxic component, isolated from the venom of Vipera ammodytes meridionalis, is the heterodimeric postsynaptic ionic complex vipoxin, composed of a basic and toxic PLA2 enzyme subunit (GIIA secreted PLA2) and an acidic, enzymatically inactive and nontoxic subunit - vipoxin acidic component (VAC). This study demonstrates for the first time that vipoxin and its individual subunits affect integrity and viability of HepG2 cells displaying differences in their pharmacological activities. Under the experimental conditions, the individual PLA2 subunit induces cytotoxicity, cytoskeletal rearrangements and triggers early apoptosis in a concentration-dependent manner related to its enzymatic activity. Vipoxin and VAC do not affect cell viability but manifest high degree of genotoxicity, whereas DNA damage induced by PLA2 subunit could be defined as moderate and not associated with its catalytic activity. Our results suggest that the interactions between vipoxin subunits play an important role in HepG2 cell response and most likely affect the observed distinction between cyto- and genotoxicity.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
3-Nitrobenzyl alcohol, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture