MilliporeSigma
  • Hybrid nanotrimers for dual T1 and T2-weighted magnetic resonance imaging.
Hybrid nanotrimers for dual T1 and T2-weighted magnetic resonance imaging.
ACS nano (2014-10-07)
Kai Cheng, Meng Yang, Ruiping Zhang, Chunxia Qin, Xinhui Su, Zhen Cheng
ABSTRACT
Development of multifunctional nanoparticle-based probes for dual T1- and T2-weighted magnetic resonance imaging (MRI) could allow us to image and diagnose the tumors or other abnormalities in an exceptionally accurate and reliable manner. In this study, by fusing distinct nanocrystals via solid-state interfaces, we built hybrid heteronanostructures to combine both T1 and T2- weighted contrast agents together for MRI with high accuracy and reliability. The resultant hybrid heterotrimers showed high stability in physiological conditions and could induce both simultaneous positive and negative contrast enhancements in MR images. Small animal positron emission tomography imaging study revealed that the hybrid heterostructures displayed favorable biodistribution and were suitable for in vivo imaging. Their potential as dual contrast agents for T1 and T2-weighted MRI was further demonstrated by in vitro and in vivo imaging and relaxivity measurements.
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleylamine, technical grade, 70%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Oleic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, technical grade, 90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Octadecene, technical grade, 90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, natural, FCC
SKU
Pack Size
Availability
Price
Quantity
Supelco
Oleic acid, Selectophore, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Oleic acid, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
SAFC
Oleylamine, ≥98% (primary amine)