MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Injury-associated PACAP expression in rat sensory and motor neurons is induced by endogenous BDNF.
Injury-associated PACAP expression in rat sensory and motor neurons is induced by endogenous BDNF.
PloS one (2014-06-27)
Lina M E Pettersson, Nicole M Geremia, Zhengxin Ying, Valerie M K Verge
ABSTRACT
Peripheral nerve injury results in dramatic upregulation in pituitary adenylate cyclase activating polypeptide (PACAP) expression in adult rat dorsal root ganglia and spinal motor neurons mirroring that described for the neurotrophin brain derived neurotrophic factor (BDNF). Thus, we posited that injury-associated alterations in BDNF expression regulate the changes in PACAP expression observed in the injured neurons. The role of endogenous BDNF in induction and/or maintenance of PACAP mRNA expression in injured adult rat motor and sensory neurons was examined by intrathecally infusing or intraperitoneally injecting BDNF-specific antibodies or control IgGs immediately at the time of L4-L6 spinal nerve injury, or in a delayed fashion one week later for 3 days followed by analysis of impact on PACAP expression. PACAP mRNA in injured lumbar sensory and motor neurons was detected using in situ hybridization, allowing quantification of relative changes between experimental groups, with ATF-3 immunofluorescence serving to identify the injured subpopulation of motor neurons. Both the incidence and level of PACAP mRNA expression were dramatically reduced in injured sensory and motor neurons in response to immediate intrathecal anti-BDNF treatment. In contrast, neither intraperitoneal injections nor delayed intrathecal infusions of anti-BDNF had any discernible impact on PACAP expression. This impact on PACAP expression in response to BDNF immunoneutralization in DRG was confirmed using qRT-PCR or by using BDNF selective siRNAs to reduce neuronal BDNF expression. Collectively, our findings support that endogenous injury-associated BDNF expression is critically involved in induction, but not maintenance, of injury-associated PACAP expression in sensory and motor neurons.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bdnf
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Thpo
Sigma-Aldrich
MISSION® esiRNA, targeting human THPO
Sigma-Aldrich
MISSION® esiRNA, targeting human BDNF