MilliporeSigma
Dapsone in the management of autoimmune bullous diseases.
Immunology and allergy clinics of North America (2012-05-09)
Evan W Piette, Victoria P Werth
ABSTRACT
Dapsone is used in the treatment of autoimmune bullous diseases (AIBD), a group of disorders resulting from autoimmunity directed against basement membrane and/or intercellular adhesion molecules on cutaneous and mucosal surfaces. This review summarizes the limited published data evaluating dapsone as a therapy for AIBD.
Sigma-Aldrich
4-Aminophenyl sulfone, 97%
Supelco
Dapsone, VETRANAL®, analytical standard
Dapsone, European Pharmacopoeia (EP) Reference Standard