MilliporeSigma
  • Rizatriptan in migraineurs with unilateral cranial autonomic symptoms: a double-blind trial.
Rizatriptan in migraineurs with unilateral cranial autonomic symptoms: a double-blind trial.
The journal of headache and pain (2012-03-31)
Piero Barbanti, Luisa Fofi, Valentina Dall'Armi, Cinzia Aurilia, Gabriella Egeo, Nicola Vanacore, Stefano Bonassi
ABSTRACT
The objective and background is to confirm in a double-blind, placebo-controlled study the high triptan response rates we had previously reported in an open study in migraine patients with unilateral cranial autonomic symptoms. In this randomized, double-blind, placebo-controlled study 80 migraineurs with unilateral cranial autonomic symptoms were assigned to receive rizatriptan 10 mg wafer or placebo (ratio 1:1) and treated for a single moderate or severe migraine attack. The primary endpoints were pain freedom at 2 h and total migraine freedom at 2 h. Secondary endpoints included pain relief, no associated symptoms and sustained pain freedom or relief. Significantly more patients reported pain freedom at 2 h after taking rizatriptan (54 %) than after placebo (8 %) (therapeutic gain 46 % [28 %; 64 %]; P < 0.001). Similarly, significantly more patients reported total migraine freedom at 2 h after rizatriptan (51 %) than after placebo (8 %) (therapeutic gain 43 % [26 %; 61 %]; P < 0.001). Rizatriptan was also more effective than placebo on most secondary endpoints. We confirm in a placebo-controlled study our previous data suggesting that the presence of unilateral cranial autonomic symptoms in migraineurs predicts a positive response to triptans, probably owing to intense trigeminal peripheral afferent activation which strongly recruits peripheral neurovascular 5-HT1B/1D receptors. Acute and preventive pharmacological trials in migraine should focus also on this subset of migraine patients.
Sigma-Aldrich
Rizatriptan benzoate salt, ≥98% (HPLC)