MilliporeSigma
  • Electron microscopical identification of 3,3',5,5'-tetramethylbenzidine-reacted horseradish peroxidase after retrograde axoplasmic transport.
Electron microscopical identification of 3,3',5,5'-tetramethylbenzidine-reacted horseradish peroxidase after retrograde axoplasmic transport.
Neuroscience letters (1981-04-09)
C Stürmer, K Bielenberg, W B Spatz
ABSTRACT
Tetramethylbenzidine (TMB) histochemistry for retrogradely transported horseradish peroxidase (HRP) has been successfully applied in electron microscopical preparations in the guinea pig visual cortex. In ultrathin sections, the TMB--HRP reaction product appears as an accumulation of typical electron-dense crystals. These crystals were identified in the cytoplasm of labeled cells in the proximal portions of the dendrites, and in myelinated axons. It is demonstrated that the TMB--HRP method is well suitable to analyze the ultrastructure of labeled cells and their synaptology.
Sigma-Aldrich
N,N,N′,N′-Tetramethylbenzidine, ≥95% (HPLC)