MilliporeSigma
  • Extravascular injection of sclerotic agents does not affect vessels in the rat: experimental implications for percutaneous sclerotherapy of arteriovenous malformations.
Extravascular injection of sclerotic agents does not affect vessels in the rat: experimental implications for percutaneous sclerotherapy of arteriovenous malformations.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2012-05-02)
D Sato, M Kurita, M Ozaki, N Kaji, A Takushima, K Harii
ABSTRACT
Sclerotherapy is useful for the treatment of arteriovenous vascular malformations. However, intravascular administration of sclerotic agents into small arteriovenous niduses is often difficult. Extravascular administration of sclerotic agents causes reduction of vascular flow on Doppler echo during clinical sclerotherapy. Therefore, we aimed to investigate whether the extravascular injection of sclerotic agents affects tiny vessels. Animal study. The effect of extravascular injection of sclerotic agents on vessels was investigated using rat femoral and superficial inferior epigastric vessels. After surgical exposure of vessels, absolute ethanol, 5% ethanolamine oleate and 3% polidocanol were injected into perivascular surrounding tissues, and their effect on vessels was evaluated after 14 days using histology and coloured silicone rubber injection. The integrity of the vascular lumen, endothelial cells and vascular patency were not affected by injection of sclerotic agents. Attenuation of vascular flow of an arteriovenous shunt after extravascular injection of sclerotic agents is transient and/or trivial and does not cause disruption of vessels. Therefore, sclerotic agents should be delivered to obtain sufficient destruction of arteriovenous malformation lesions and blood flow.
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
Thesit®
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Brij® L4, average Mn ~362
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether