MilliporeSigma
  • Methyl ethyl ketone peroxide ingestion: a rare cause of corrosive chemical poisoning.
Methyl ethyl ketone peroxide ingestion: a rare cause of corrosive chemical poisoning.
The National medical journal of India (2010-10-19)
M V S Subbalaxmi, S Abkari, V R Srinivasan, A Krishnaprasad
ABSTRACT
Methyl ethyl ketone peroxide, commonly used in the lamination industry, is a highly inflammable substance. Ingestion of methyl ethyl ketone peroxide is associated with a high morbidity and mortality. It can cause injury to the gastrointestinal tract on ingestion, leading to gangrene or perforation. Gastric lavage is contraindicated as it is a highly corrosive substance. We report a patient with poisoning due to methyl ethyl ketone peroxide who presented with upper gastrointestinal bleeding, and developed gangrene of the bowel and died of septicaemia and multiorgan failure.
Sigma-Aldrich
Luperox® DDM-9, 2-Butanone peroxide solution, ~35 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate
Sigma-Aldrich
Luperox® DHD-9, 2-Butanone peroxide solution, ~32 wt. % in phthalate-free plasticizer mixture