MilliporeSigma
  • New cyclodextrin derivative 6-O-(2-hydroxybutyl)-beta-cyclodextrin: preparation and its application in molecular binding and recognition.
New cyclodextrin derivative 6-O-(2-hydroxybutyl)-beta-cyclodextrin: preparation and its application in molecular binding and recognition.
Carbohydrate research (2009-08-26)
Hong Yuan Sun, Yan Bai, Ming Gang Zhao, Ai You Hao, Gui Ying Xu, Jian Shen, Jian Ye Li, Tao Sun, Hua Cheng Zhang
ABSTRACT
A new soluble cyclodextrin derivative 6-O-(2-hydroxybutyl)-beta-cyclodextrin (6-HB-beta-CD) was prepared. Its molecular binding and recognition ability were investigated with the comparison of beta-cyclodextrin (beta-CD), 2-O-(2-hydroxypropyl)-beta-cyclodextrin (2-HP-beta-CD), 6-O-(2-hydroxypropyl)-beta-cyclodextrin (6-HP-beta-CD), and 2-O-(2-hydroxybutyl)-beta-cyclodextrin (2-HB-beta-CD). The relationship between the complex stability constants and the possible structures of inclusion compounds was discussed with the interaction of hosts and guests, including the weak hydrophobic interactions, the size/shape matching, the steric hindrance, and the hydrophilic property.
Sigma-Aldrich
1,2-Epoxybutane, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-(−)-1,2-Epoxybutane, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
(R)-(+)-1,2-Epoxybutane, 98%
Pricing and availability is not currently available.