MilliporeSigma
Ultrasound assisted synthesis of nano-sized lithium cobalt oxide.
Ultrasonics sonochemistry (2008-05-09)
Kwang-Heon Kim, Kwang-Bum Kim
ABSTRACT
Nano-sized HT-LiCoO(2) powders were prepared by sonochemical synthesis in an aqueous solution of lithium hydroxide containing cobalt hydroxide at approximately 80 degrees C without any further heat treatment at high temperature. The effects of the LiOH concentration, oxidation conditions, ultrasound irradiation time and temperature on the formation of the nano-sized HT-LiCoO(2) phase were investigated. The formation of the HT-LiCoO(2) phase was confirmed by X-ray diffraction and Raman spectroscopy. The TEM images showed the presence of HT-LiCoO(2) aggregates with a mean particle diameter of approximately 20 nm. The reaction mechanism of the ultrasound assisted synthesis of nano-sized LiCoO(2) was proposed on the basis of the XRD, X-ray absorption spectroscopy analysis and TEM observation of the reaction products taken during the course of the synthesis.
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, purum, ≥98.5%
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%