MilliporeSigma
  • Plasma pharmacokinetics of (poly)phenol metabolites and catabolites after ingestion of orange juice by endurance trained men.
Plasma pharmacokinetics of (poly)phenol metabolites and catabolites after ingestion of orange juice by endurance trained men.
Free radical biology & medicine (2020-09-15)
Gema Pereira-Caro, Michael N Clifford, Thelma Polyviou, Iziar A Ludwig, Hani Alfheeaid, José Manuel Moreno-Rojas, Ada L Garcia, Dalia Malkova, Alan Crozier
ABSTRACT
The health benefits of orange juice (OJ) consumption are attributed in part to the circulating flavanone phase II metabolites and their microbial-derived ring fission phenolic catabolites. The present study investigated these compounds in the bloodstream after acute intake of 500 mL of OJ. Plasma samples obtained at 0, 1, 2, 3, 4, 5, 6, 7, 8 and 24 h after OJ intake were analysed by HPLC-HR-MS. Eleven flavanone metabolites and 36 phenolic catabolites were identified and quantified in plasma. The main metabolites were hesperetin-3'-sulfate with a peak plasma concentration (Cmax) of 80 nmol/L, followed by hesperetin-7-glucuronide (Cmax 24 nmol/L), hesperetin-3'-glucuronide (Cmax 18 nmol/L) and naringenin-7-glucuronide (Cmax 21 nmol/L). Among the main phenolic catabolites to increase in plasma after OJ consumption were 3'-methoxycinnamic acid-4'-sulfate (Cmax 19 nmol/L), 3-hydroxy-3-(3'-hydroxy-4'-methoxyphenyl)propanoic acid (Cmax 20 nmol/L), 3-(3'-hydroxy-4'-methoxyphenyl)propanoic acid (Cmax 19 nmol/L), 3-(4'-hydroxyphenyl)propanoic acid (Cmax 25 nmol/L), and 3-(phenyl)propanoic acid (Cmax 19 nmol/L), as well as substantial amounts of phenylacetic and hippuric acids. The comprehensive plasma pharmacokinetic profiles that were obtained are of value to the design of future ex vivo cell studies, aimed at elucidating the mechanisms underlying the potential health benefits of OJ consumption. CLINICAL TRIAL REGISTRATION NUMBER: This trial was registered at clinicaltrials.gov as NCT02627547.
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, ≥96.0% (T)
Supelco
3-(3-Hydroxyphenyl)-3-hydroxypropionic acid, analytical standard