MilliporeSigma
  • Ixorapeptide I and ixorapeptide II, bioactive peptides isolated from Ixora coccinea.
Ixorapeptide I and ixorapeptide II, bioactive peptides isolated from Ixora coccinea.
Bioorganic & medicinal chemistry letters (2010-11-26)
Chia-Lin Lee, Yung-Chih Liao, Tsong-Long Hwang, Chin-Chung Wu, Fang-Rong Chang, Yang-Chang Wu
ABSTRACT
Two novel derivatized peptides, designated as ixorapeptide I (1) and ixorapeptide II (2), in addition to 28 other known compounds, were isolated from the MeOH extract of Ixora coccinea using bioassay-guided fractionation. The structures of metabolites 1 and 2 were determined by interpretation of the spectroscopic data and Marfey's method. Compound 1 exhibited selective potency against Hep3B liver cancer cell line with an IC(50) value of 3.36 μg/mL, and compound 2 did not show notable cytotoxicity toward cancer cell lines but could inhibit superoxide anion generation and elastase release with IC(50) values of 0.21 and 0.27 μg/mL, respectively. Moreover, kaempferol and luteolin from this plant showed inhibition with IC(50) values of 3.55 and 2.56 μg/mL, respectively on platelet aggregation induced by collagen.
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Supelco
Ursolic acid, analytical standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Ursolic acid, ≥90%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Scopoletin, ≥99%
Sigma-Aldrich
Lupeol, ≥94%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)