MilliporeSigma
  • Novel UBQLN2 mutations linked to amyotrophic lateral sclerosis and atypical hereditary spastic paraplegia phenotype through defective HSP70-mediated proteolysis.
Novel UBQLN2 mutations linked to amyotrophic lateral sclerosis and atypical hereditary spastic paraplegia phenotype through defective HSP70-mediated proteolysis.
Neurobiology of aging (2017-07-19)
Elisa Teyssou, Laura Chartier, Maria-Del-Mar Amador, Roselina Lam, Géraldine Lautrette, Marie Nicol, Selma Machat, Sandra Da Barroca, Carine Moigneu, Mathilde Mairey, Thierry Larmonier, Safaa Saker, Christelle Dussert, Sylvie Forlani, Bertrand Fontaine, Danielle Seilhean, Delphine Bohl, Séverine Boillée, Vincent Meininger, Philippe Couratier, François Salachas, Giovanni Stevanin, Stéphanie Millecamps
ABSTRACT
Mutations in UBQLN2 have been associated with rare cases of X-linked juvenile and adult forms of amyotrophic lateral sclerosis (ALS) and ALS linked to frontotemporal dementia (FTD). Here, we report 1 known (c.1489C>T, p.Pro497Ser, P497S) and 3 novel (c.1481C>T, p.Pro494Leu, P494L; c.1498C>T, p.Pro500Ser, P500S; and c.1516C>G, p.Pro506Ala, P506A) missense mutations in the PXX domain of UBQLN2 in familial motor neuron diseases including ALS and spastic paraplegia (SP). A novel missense mutation (c.1462G>A, p.Ala488Thr, A488T) adjacent to this hotspot UBQLN2 domain was identified in a sporadic case of ALS. These mutations are conserved in mammals, are absent from ExAC and gnomAD browsers, and are predicted to be deleterious by SIFT in silico analysis. Patient lymphoblasts carrying a UBQLN2 mutation showed absence of ubiquilin-2 accumulation, disrupted binding with HSP70, and impaired autophagic pathway. Our results confirm the role of PXX repeat in ALS pathogenesis, show that UBQLN2-linked disease can manifest like a SP phenotype, evidence a highly reduced disease penetrance in females carrying UBQLN2 mutations, which is important information for genetic counseling, and underline the pivotal role of ubiquilin-2 in proteolysis regulation pathways.
Sigma-Aldrich
Anti-p62 (Sequestosome-1) Antibody, clone 11C9.2, clone 11C9.2, from mouse
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Heat Shock Protein 70 Antibody, clone W27, clone W27, Chemicon®, from mouse