MilliporeSigma
Bioactive constituents from Boswellia papyrifera.
Journal of natural products (2005-02-26)
Atta-ur-Rahman, Humera Naz, Fadimatou, Talat Makhmoor, Amsha Yasin, Naheed Fatima, F N Ngounou, S F Kimbu, B L Sondengam, M Iqbal Choudhary
ABSTRACT
Phytochemical investigation of the stem bark extract of Boswellia papyrifera afforded two new stilbene glycosides, trans-4',5-dihydroxy-3-methoxystilbene-5-O-{alpha-L-rhamnopyranosyl-(1-->2)-[alpha-L-rhamnopyranosyl-(1-->6)]-beta-D-glucopyranoside (1), trans-4',5-dihydroxy-3-methoxystilbene-5-O-[alpha-L-rhamnopyranosyl-(1-->6)]-beta-D-glucopyranoside (2), and a new triterpene, 3alpha-acetoxy-27-hydroxylup-20(29)-en-24-oic acid (3), along with five known compounds, 11-keto-beta-boswellic acid (4), beta-elemonic acid (7), 3alpha-acetoxy-11-keto-beta-boswellic acid (8), beta-boswellic acid (9), and beta-sitosterol (10). The stilbene glycosides exhibited significant inhibition of phosphodiesterase I and xanthine oxidase. The triterpenes (3-9) exhibited prolyl endopeptidase inhibitory activities.
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)